Assay Details
| Gene Symbol: CCRL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075732.2 | NP_001069200.1 | 1 - 2 | 232 | 61 | |
| GenBank mRNA | BC122767.1 | - | 1 - 2 | 234 | 61 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC122767.1, NM_001075732.2 transcripts.
Set Membership: |
|

CCRL2
Genomic Map