Assay Details
| Gene Symbol: GSTT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075738.1 | NP_001069206.1 | 4 - 5 | 597 | 66 | |
| GenBank mRNA | BC120024.1 | - | 4 - 5 | 597 | 66 | |
More Information
Set Membership: |
|

GSTT2
Genomic Map