Assay Details
| Gene Symbol: CTCF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075748.1 | NP_001069216.1 | 10 - 11 | 2062 | 66 | |
| GenBank mRNA | BC123741.1 | - | 10 - 11 | 2062 | 66 | |
More Information
Set Membership: |
|

CTCF
Genomic Map