Assay Details
| Gene Symbol: KLC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075768.1 | NP_001069236.1 | 9 - 10 | 1346 | 68 | |
| GenBank mRNA | BC123748.1 | - | 9 - 10 | 1346 | 68 | |
More Information
Set Membership: |
|

KLC2
Genomic Map