Assay Details
| Gene Symbol: OSTM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075776.1 | NP_001069244.1 | 5 - 6 | 1051 | 65 | |
| GenBank mRNA | BC120014.1 | - | 5 - 6 | 1051 | 65 | |
More Information
Set Membership: |
|

OSTM1
Genomic Map