Assay Details
| Gene Symbol: DLL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075779.1 | NP_001069247.1 | 8 - 9 | 1518 | 64 | |
| GenBank mRNA | BC140546.1 | - | 8 - 9 | 1445 | 64 | |
| BT026552.1 | - | 8 - 9 | 1518 | 64 | ||
More Information
Set Membership: |
|

DLL4
Genomic Map