Assay Details
| Gene Symbol: TTLL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075793.2 | NP_001069261.1 | 1 - 2 | 284 | 136 | |
| GenBank mRNA | BC122707.1 | - | 1 - 2 | 285 | 136 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC122707.1, NM_001075793.2 transcripts.
Set Membership: |
|

TTLL2
Genomic Map