Assay Details
| Gene Symbol: RDH16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075796.1 | NP_001069264.1 | 2 - 3 | 578 | 97 | |
| GenBank mRNA | BC151534.1 | - | 2 - 3 | 550 | 97 | |
| BT026289.1 | - | 2 - 3 | 578 | 97 | ||
More Information
Set Membership: |
|

RDH16
Genomic Map