Assay Details
| Gene Symbol: KRT27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075815.1 | NP_001069283.1 | 1 - 2 | 479 | 67 | |
| GenBank mRNA | BC119954.1 | - | 1 - 2 | 479 | 67 | |
More Information
Set Membership: |
|

KRT27
Genomic Map