Assay Details
| Gene Symbol: NMNAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075834.1 | NP_001069302.1 | 4 - 5 | 532 | 80 | |
| GenBank mRNA | BC119834.1 | - | 4 - 5 | 532 | 80 | |
More Information
Set Membership: |
|

NMNAT1
Genomic Map