Assay Details
| Gene Symbol: NKIRAS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075835.1 | NP_001069303.1 | 4 - 5 | 1271 | 64 | |
| GenBank mRNA | BC120341.1 | - | 4 - 5 | 1271 | 64 | |
More Information
Set Membership: |
|

NKIRAS1
Genomic Map