Assay Details
| Gene Symbol: GAL3ST3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075840.1 | NP_001069308.1 | 1 - 2 | 206 | 107 | |
| GenBank mRNA | BC120166.1 | - | 1 - 2 | 206 | 107 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC120166.1, NM_001075840.1 transcripts.
Set Membership: |
|

GAL3ST3
Genomic Map