Assay Details
| Gene Symbol: ADH6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075843.1 | NP_001069311.1 | 8 - 9 | 1149 | 147 | |
| GenBank mRNA | BC120378.1 | - | 8 - 9 | 1149 | 147 | |
More Information
Set Membership: |
|

ADH6
Genomic Map