Assay Details
| Gene Symbol: PPP2CB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075857.1 | NP_001069325.1 | 5 - 6 | 749 | 65 | |
| GenBank mRNA | BC120332.1 | - | 5 - 6 | 749 | 65 | |
More Information
Set Membership: |
|

PPP2CB
Genomic Map