Assay Details
| Gene Symbol: SERPINB9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075859.2 | NP_001069327.1 | 1 - 2 | 35 | 61 | |
| GenBank mRNA | BC123622.1 | - | 1 - 2 | 36 | 61 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC123622.1, NM_001075859.2 transcripts.
Set Membership: |
|

SERPINB9
Genomic Map