Assay Details
| Gene Symbol: SERPINB9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075859.2 | NP_001069327.1 | 3 - 4 | 307 | 136 | |
| GenBank mRNA | BC123622.1 | - | 3 - 4 | 308 | 136 | |
More Information
Set Membership: |
|

SERPINB9
Genomic Map