Assay Details
| Gene Symbol: MMD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075870.2 | NP_001069338.1 | 3 - 4 | 343 | 64 | |
| GenBank mRNA | BC120183.1 | - | 3 - 4 | 344 | 64 | |
More Information
Set Membership: |
|

MMD2
Genomic Map