Assay Details
| Gene Symbol: LNX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075884.1 | NP_001069352.1 | 8 - 9 | 2038 | 66 | |
| GenBank mRNA | BC119977.1 | - | 8 - 9 | 2038 | 66 | |
More Information
Set Membership: |
|

LNX2
Genomic Map