Assay Details
| Gene Symbol: NR4A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075911.1 | NP_001069379.1 | 4 - 5 | 1227 | 51 | |
| GenBank mRNA | BT026269.1 | - | 4 - 5 | 1227 | 51 | |
More Information
Set Membership: |
|

NR4A1
Genomic Map