Assay Details
| Gene Symbol: KRT85 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075913.1 | NP_001069381.1 | 8 - 9 | 1387 | 86 | |
| GenBank mRNA | BC119906.1 | - | 8 - 9 | 1387 | 86 | |
More Information
Set Membership: |
|

KRT85
Genomic Map