Assay Details
| Gene Symbol: MAPK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075920.2 | NP_001069388.1 | 1 - 2 | 608 | 68 | |
| GenBank mRNA | BC122768.1 | - | 1 - 2 | 609 | 68 | |
More Information
Set Membership: |
|

MAPK4
Genomic Map