Assay Details
| Gene Symbol: SDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075924.1 | NP_001069392.1 | 2 - 3 | 366 | 68 | |
| GenBank mRNA | BC123495.1 | - | 2 - 3 | 366 | 68 | |
More Information
Set Membership: |
|

SDC1
Genomic Map