Assay Details
| Gene Symbol: AGPAT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075932.1 | NP_001069400.1 | 7 - 8 | 1006 | 77 | |
| GenBank mRNA | BC122693.1 | - | 7 - 8 | 1006 | 77 | |
More Information
Set Membership: |
|

AGPAT5
Genomic Map