Assay Details
| Gene Symbol: KRT72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075944.1 | NP_001069412.1 | 8 - 9 | 1401 | 69 | |
| GenBank mRNA | BC118311.1 | - | 8 - 9 | 1401 | 69 | |
More Information
Set Membership: |
|

KRT72
Genomic Map