Assay Details
| Gene Symbol: CALY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075953.2 | NP_001069421.1 | 4 - 5 | 416 | 72 | |
| GenBank mRNA | BC120132.1 | - | 4 - 5 | 417 | 72 | |
More Information
Set Membership: |
|

CALY
Genomic Map