Assay Details
| Gene Symbol: KRT71 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075970.1 | NP_001069438.1 | 6 - 7 | 1166 | 85 | |
| GenBank mRNA | BC118317.1 | - | 6 - 7 | 1166 | 85 | |
More Information
Set Membership: |
|

KRT71
Genomic Map