Assay Details
| Gene Symbol: KPTN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076005.1 | NP_001069473.1 | 4 - 5 | 506 | 72 | |
| GenBank mRNA | BC122686.1 | - | 4 - 5 | 506 | 72 | |
More Information
Set Membership: |
|

KPTN
Genomic Map