Assay Details
| Gene Symbol: FGF18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076007.2 | NP_001069475.1 | 1 - 2 | 149 | 83 | |
| GenBank mRNA | BC120280.1 | - | 1 - 2 | 150 | 83 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC120280.1, NM_001076007.2 transcripts.
Set Membership: |
|

FGF18
Genomic Map