Assay Details
| Gene Symbol: MSANTD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076019.1 | NP_001069487.1 | 2 - 3 | 489 | 83 | |
| GenBank mRNA | BC122633.1 | - | 2 - 3 | 489 | 83 | |
More Information
Set Membership: |
|

MSANTD3
Genomic Map