Assay Details
| Gene Symbol: DNAAF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076024.3 | NP_001069492.2 | 2 - 3 | 2213 | 131 | |
| GenBank mRNA | BC120319.1 | - | 2 - 3 | 767 | 131 | |
More Information
Set Membership: |
|

DNAAF2
Genomic Map