Assay Details
| Gene Symbol: NMRAL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076032.2 | NP_001069500.1 | 4 - 5 | 608 | 75 | |
| GenBank mRNA | BC120089.1 | - | 4 - 5 | 609 | 75 | |
More Information
Set Membership: |
|

NMRAL1
Genomic Map