Assay Details
| Gene Symbol: KRT35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076073.1 | NP_001069541.1 | 6 - 7 | 1246 | 127 | |
| GenBank mRNA | BC119951.1 | - | 6 - 7 | 1246 | 127 | |
More Information
Set Membership: |
|

KRT35
Genomic Map