Assay Details
| Gene Symbol: PAPSS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076075.2 | NP_001069543.1 | 11 - 12 | 1802 | 93 | |
| GenBank mRNA | BC120295.1 | - | 11 - 12 | 1803 | 93 | |
More Information
Set Membership: |
|

PAPSS2
Genomic Map