Assay Details
| Gene Symbol: PMM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076083.1 | NP_001069551.1 | 7 - 8 | 1018 | 71 | |
| GenBank mRNA | BC120394.1 | - | 7 - 8 | 1018 | 71 | |
More Information
Set Membership: |
|

PMM1
Genomic Map