Assay Details
| Gene Symbol: CREBL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076088.1 | NP_001069556.1 | 2 - 3 | 433 | 85 | |
| GenBank mRNA | BC119861.1 | - | 2 - 3 | 433 | 85 | |
More Information
Set Membership: |
|

CREBL2
Genomic Map