Assay Details
| Gene Symbol: SULT1C4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076097.1 | NP_001069565.1 | 3 - 4 | 485 | 174 | |
| GenBank mRNA | BC109942.1 | - | 3 - 4 | 485 | 174 | |
More Information
Set Membership: |
|

SULT1C4
Genomic Map