Assay Details
| Gene Symbol: MINDY3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076107.1 | NP_001069575.1 | 7 - 8 | 765 | 128 | |
| GenBank mRNA | BC122636.1 | - | 7 - 8 | 765 | 128 | |
More Information
Set Membership: |
|

MINDY3
Genomic Map