Assay Details
| Gene Symbol: ARL4D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076135.1 | NP_001069603.1 | 1 - 2 | 63 | 106 | |
| GenBank mRNA | BC120393.1 | - | 1 - 2 | 63 | 106 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC120393.1, NM_001076135.1 transcripts.
Set Membership: |
|

ARL4D
Genomic Map