Assay Details
| Gene Symbol: IL1RAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076155.1 | NP_001069623.1 | 5 - 6 | 841 | 60 | |
| GenBank mRNA | BC120353.1 | - | 5 - 6 | 841 | 60 | |
More Information
Set Membership: |
|

IL1RAP
Genomic Map