Assay Details
Gene Symbol: C1GALT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076160.2 | NP_001069628.1 | 3 - 4 | 1008 | 146 | |
GenBank mRNA | BC120299.1 | - | 3 - 4 | 1009 | 146 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
