Assay Details
| Gene Symbol: PPP1R3C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076164.2 | NP_001069632.1 | 1 - 2 | 105 | 59 | |
| GenBank mRNA | BC120042.1 | - | 1 - 2 | 107 | 59 | |
| BT030698.1 | - | 1 - 2 | 70 | 59 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC120042.1, NM_001076164.2 transcripts.
Set Membership: |
|

PPP1R3C
Genomic Map