Assay Details
| Gene Symbol: FN3K | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076182.1 | NP_001069650.1 | 2 - 3 | 397 | 78 | |
| GenBank mRNA | BC120409.1 | - | 2 - 3 | 397 | 78 | |
More Information
Set Membership: |
|

FN3K
Genomic Map