Assay Details
| Gene Symbol: NPTN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076199.1 | NP_001069667.1 | 4 - 5 | 627 | 61 | |
| GenBank mRNA | BC119903.1 | - | 4 - 5 | 627 | 61 | |
More Information
Set Membership: |
|

NPTN
Genomic Map