Assay Details
| Gene Symbol: FBXO25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076211.1 | NP_001069679.1 | 2 - 3 | 247 | 86 | |
| GenBank mRNA | BC114733.1 | - | 2 - 3 | 247 | 86 | |
More Information
Set Membership: |
|

FBXO25
Genomic Map