Assay Details
| Gene Symbol: GPR162 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076217.1 | NP_001069685.1 | 2 - 3 | 1345 | 72 | |
| GenBank mRNA | BC119846.1 | - | 2 - 3 | 1345 | 72 | |
More Information
Set Membership: |
|

GPR162
Genomic Map