Assay Details
| Gene Symbol: SOCS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076218.2 | NP_001069686.1 | 1 - 2 | 245 | 127 | |
| GenBank mRNA | BC120292.1 | - | 1 - 2 | 246 | 127 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC120292.1, NM_001076218.2 transcripts.
Set Membership: |
|

SOCS4
Genomic Map