Assay Details
| Gene Symbol: ABI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076241.1 | NP_001069709.1 | 7 - 8 | 944 | 55 | |
| GenBank mRNA | BC119969.1 | - | 7 - 8 | 944 | 55 | |
More Information
Set Membership: |
|

ABI1
Genomic Map