Assay Details
| Gene Symbol: MAGOHB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076252.1 | NP_001069720.1 | 1 - 2 | 153 | 95 | |
| GenBank mRNA | BC120291.1 | - | 1 - 2 | 153 | 95 | |
More Information
Set Membership: |
|

MAGOHB
Genomic Map