Assay Details
| Gene Symbol: MXD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076256.1 | NP_001069724.1 | 4 - 5 | 453 | 93 | |
| GenBank mRNA | BC120230.1 | - | 4 - 5 | 453 | 93 | |
| BT030600.2 | - | 4 - 5 | 347 | 93 | ||
More Information
Set Membership: |
|

MXD4
Genomic Map