Assay Details
Gene Symbol: PTX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076259.2 | NP_001069727.1 | 1 - 2 | 219 | 142 | |
GenBank mRNA | BC120175.1 | - | 1 - 2 | 220 | 142 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
